Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl oxazole-4,5-dicarboxylate, 99%

$89.91 - $561.84

Chemical Identifiers

Chemical Name or Material Dimethyl oxazole-4,5-dicarboxylate
CAS 72030-81-0
Molecular Formula C7H7NO5
Formula Weight 185.14
MDL Number MFCD05863258
Synonym Oxazole-4,5-dicarboxylic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3000903

 
alfa aesar™
H3000903
1g Each for $89.91

AAH3000909

 
alfa aesar™
H3000909
10g Each for $561.84
Description & Specifications

Specifications

Formula Weight 185.14
Chemical Name or Material Dimethyl oxazole-4,5-dicarboxylate
Assay 99%
Molecular Formula C7H7NO5
Melting Point 74° to 76°C
Boiling Point 150°C (12mmHg)
MDL Number MFCD05863258
CAS 72030-81-0
Quantity 1g
Synonym Oxazole-4,5-dicarboxylic acid dimethyl ester

Applications
The teroxazole has been synthesized from dimethyl oxazole-4,5-dicarboxylate and lithiobenzyl isocyanide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.