missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl oxalate, 99%

Manufacturer:  Alfa Aesar™ A1535922

 View more versions of this product

Catalog No. AAA1535922



Description

Description

Applications
Dimethyl oxalate is used as an alternate fuel in the direct oxidation fuel cell. It is involved in the catalytic hydrogenation using Cu/SiO2 to prepare ethylene glycol. It is also used in the selective gas-phase hydrogenation catalyzed by Ag/SiO2 to get the corresponding alcohol.

Solubility
Soluble in ethanol. Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
Specifications

Specifications

Dimethyl oxalate
162°C to 163°C
100g
C4H6O4
1071744
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid
COC(=O)C(=O)OC
118.088
CHEBI:6859
99%
75°C (167°F)
50°C to 54°C
553-90-2
MFCD00008442
14,6106
LOMVENUNSWAXEN-UHFFFAOYSA-N
dimethyl oxalate
11120
118.09

Chemical Identifiers

553-90-2
118.088
LOMVENUNSWAXEN-UHFFFAOYSA-N
11120
dimethyl oxalate
C4H6O4
MFCD00008442
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid
CHEBI:6859
COC(=O)C(=O)OC
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RO2850000

TSCA : TSCA

Documents