missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl octafluoroadipate, 94%

$77.73 - $302.26

Chemical Identifiers

CAS 3107-98-0
Molecular Formula C8H6F8O4
Molecular Weight (g/mol) 318.119
MDL Number MFCD00042267
InChI Key XPXVIIILXUOEQA-UHFFFAOYSA-N
Synonym dimethyl octafluoroadipate, dimethyl perfluoroadipate, hexanedioic acid, octafluoro-, dimethyl ester, 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate, dimethyloctafluoroadipate, acmc-1cson, dimethyl octafluoroadipinate, dimethyl octafluorohexanedioate, perfluoroadipic acid dimethyl ester, octafluoroadipic acid dimethyl ester
PubChem CID 2737117
IUPAC Name dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate
SMILES COC(=O)C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1675206 View Documents Alfa Aesar™
L1675206
5g Each for $77.73
 
AAL1675214 View Documents Alfa Aesar™
L1675214
25g Each for $302.26
 
Specifications

Chemical Identifiers

3107-98-0
318.119
XPXVIIILXUOEQA-UHFFFAOYSA-N
2737117
COC(=O)C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F
C8H6F8O4
MFCD00042267
dimethyl octafluoroadipate, dimethyl perfluoroadipate, hexanedioic acid, octafluoro-, dimethyl ester, 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate, dimethyloctafluoroadipate, acmc-1cson, dimethyl octafluoroadipinate, dimethyl octafluorohexanedioate, perfluoroadipic acid dimethyl ester, octafluoroadipic acid dimethyl ester
dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate

Specifications

120°C (15mmHg)
3107-98-0
CH3O2C(CF2)4CO2CH3
1916112
XPXVIIILXUOEQA-UHFFFAOYSA-N
dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate
2737117
94%
5g
C8H6F8O4
MFCD00042267
dimethyl octafluoroadipate, dimethyl perfluoroadipate, hexanedioic acid, octafluoro-, dimethyl ester, 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate, dimethyloctafluoroadipate, acmc-1cson, dimethyl octafluoroadipinate, dimethyl octafluorohexanedioate, perfluoroadipic acid dimethyl ester, octafluoroadipic acid dimethyl ester
COC(=O)C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F
318.119
318.12
Dimethyl octafluoroadipate