Please login to your online account to display your discounted pricing

Dimethyl nitroterephthalate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dimethyl nitroterephthalate
Name Note 99%
CAS 5292-45-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10 H9 N O6
Formula Weight 239.18
MDL Number MFCD00008422
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194772500

  SDS
thermo fisher scientific
194772500
Plastic bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 239.18
Quantity 250g
Color Light yellow
TSCA TSCA
EINECS Number 226-134-1
Melting Point 73° to 76°C
Beilstein 09, 826
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 99%
MDL Number MFCD00008422
Chemical Name or Material Dimethyl nitroterephthalate
CAS 5292-45-5
Name Note 99%
Molecular Formula C10 H9 N O6
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.