missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl N,N-diisopropylphosphoramidite

$120.69 - $515.66

Chemical Identifiers

CAS 122194-07-4
Molecular Formula C8H20NO2P
Linear Formula (CH3O)2PN[CH(CH3)2]2
Formula Weight 193.23
MDL Number MFCD00153505
Chemical Name or Material Dimethyl N,N-diisopropylphosphoramidite
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2772303 SDS Alfa Aesar™
H2772303
1g Each for $120.69
AAH2772306 SDS Alfa Aesar™
H2772306
5g Each for $515.66
Specifications Description & Specifications

Chemical Identifiers

122194-07-4
(CH3O)2PN[CH(CH3)2]2
MFCD00153505
C8H20NO2P
193.23
Dimethyl N,N-diisopropylphosphoramidite

Specifications

122194-07-4
(CH3O)2PN[CH(CH3)2]2
MFCD00153505
Dimethyl N,N-diisopropylphosphoramidite
C8H20NO2P
193.23
1g