missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α,α-Dimethyl-β-methylsuccinimide, 99+%, ACROS Organics™

Chemical Identifiers

CAS 61748-86-5
Assay Percent Range 99+%
Molecular Formula C7H11NO2
Formula Weight 141.17
MDL Number MFCD00005497
Chemical Name or Material α, α-Dimethyl-β-methylsuccinimide
Assay ≥99.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC172512500 SDS Thermo Fisher Scientific
172512500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

61748-86-5
C7H11NO2
MFCD00005497
≥99.0%
99+%
141.17
α, α-Dimethyl-β-methylsuccinimide

Specifications

Crystalline Powder
250mg
117° to 122°C
Glass bottle
99+%
C7H11NO2
α, α-Dimethyl-β-methylsuccinimide
141.17
White
21, 394
Authentic
MFCD00005497
61748-86-5
≥99.0%

Safety and Handling

EINECSNumber : 262-956-7

TSCA : TSCA