Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl methylphosphonate, 97%

Chemical Identifiers

Chemical Name or Material Dimethyl methylphosphonate
CAS 756-79-6
Molecular Formula C3H9O3P
Formula Weight 124.08
MDL Number MFCD00008349
Beilstein 878263
Merck Index 14,3391
Synonym DMMP; Methylphosphonic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1426822

 
alfa aesar™
A1426822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dimethyl methylphosphonate
CAS 756-79-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H340-H319-H227
May cause genetic defects.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P281-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SZ9120000
TSCA TSCA
Assay 97%
Molecular Formula C3H9O3P
Boiling Point 180° to 181°C
Formula Weight 124.08
MDL Number MFCD00008349
Beilstein 878263
Quantity 100g
Flash Point 68°C (154°F)
Merck Index 14,3391
Synonym DMMP; Methylphosphonic acid dimethyl ester

Applications
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

Solubility
Miscible with water, ethanol, ether, alcohol, benzene, acetone and carbon tetrachloride. Immiscible with heavy mineral oil.

Notes
Incompatible materials strong oxidizing agents and strong bases.