missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl Methoxymethylenemalonate 98.0+%, TCI America™

$59.25 - $577.70

Chemical Identifiers

CAS 22398-14-7
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.152
MDL Number MFCD00038225
InChI Key RHFZTBSULNJWEI-UHFFFAOYSA-N
Synonym 1,3-dimethyl 2-methoxymethylidene propanedioate
PubChem CID 90770
IUPAC Name dimethyl 2-(methoxymethylidene)propanedioate
SMILES COC=C(C(=O)OC)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M120725G View Documents Tci America
M120725G
25g Each for $59.25
 
M1207500G View Documents Tci America
M1207500G
500g Each for $577.70
 
Specifications

Chemical Identifiers

22398-14-7
174.152
RHFZTBSULNJWEI-UHFFFAOYSA-N
90770
COC=C(C(=O)OC)C(=O)OC
C7H10O5
MFCD00038225
1,3-dimethyl 2-methoxymethylidene propanedioate
dimethyl 2-(methoxymethylidene)propanedioate

Specifications

167°C
43°C
2.2 (190g/L H2O soln. 20°C)
C7H10O5
1,3-dimethyl 2-methoxymethylidene propanedioate
COC=C(C(=O)OC)C(=O)OC
174.152
174.15
≥98.0% (GC)
Undesignated
25g
22398-14-7
MFCD00038225
RHFZTBSULNJWEI-UHFFFAOYSA-N
dimethyl 2-(methoxymethylidene)propanedioate
90770
Crystal-Powder at 20°C
Dimethyl Methoxymethylenemalonate
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0676

TSCA : No