missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl methoxymethylenemalonate, 98+%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 269560250

 View more versions of this product

Catalog No. AC269560250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Dimethyl methoxymethylenemalonate
22398-14-7
40°C to 44°C
158°C
Authentic
CH3OCH=C(COOCH3)2
Glass bottle
25g
RHFZTBSULNJWEI-UHFFFAOYSA-N
dimethyl 2-(methoxymethylidene)propanedioate
90770
Crystalline Solid
98+%
White
98°C (0.5mmHg)
98% min. (GC)
C7H10O5
MFCD00038225
1,3-dimethyl 2-methoxymethylidene propanedioate, acmc-1cq9e, dimethyl 2-methoxymethylene malonate, dimethyl methoxymethylene malonate, dimethyl methoxymethylenemalonate, dimethyl methoxymethylidenemalonate, dimethylmethoxymethylenemalonate, methoxymethylenemalonic acid dimethyl ester, propanedioic acid, methoxymethylene-, dimethyl ester
Solubility in water: may hydrolyse. Other solubilities: soluble in methanol 1260g/L (20 c)
COC=C(C(=O)OC)C(=O)OC
174.16
174.16
98+%

Chemical Identifiers

22398-14-7
174.16
RHFZTBSULNJWEI-UHFFFAOYSA-N
90770
COC=C(C(=O)OC)C(=O)OC
C7H10O5
MFCD00038225
1,3-dimethyl 2-methoxymethylidene propanedioate, acmc-1cq9e, dimethyl 2-methoxymethylene malonate, dimethyl methoxymethylene malonate, dimethyl methoxymethylenemalonate, dimethyl methoxymethylidenemalonate, dimethylmethoxymethylenemalonate, methoxymethylenemalonic acid dimethyl ester, propanedioic acid, methoxymethylene-, dimethyl ester
dimethyl 2-(methoxymethylidene)propanedioate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-957-4

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only