Please login to your online account to display your discounted pricing

Dimethyl methoxymethylenemalonate, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dimethyl methoxymethylenemalonate
Name Note 98+%
CAS 22398-14-7
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H10O5
Linear Formula CH3OCH=C(COOCH3)2
Formula Weight 174.16
MDL Number MFCD00038225
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC269560250

  SDS
fisher scientific acros
269560250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC269562500

  SDS
fisher scientific acros
269562500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Crystalline Solid
Formula Weight 174.16
Quantity 25g
Color White
EINECS Number 244-957-4
Melting Point 40° to 44°C
Chemical Name or Material Dimethyl methoxymethylenemalonate
Packaging Glass bottle
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C7H10O5
Linear Formula CH3OCH=C(COOCH3)2
MDL Number MFCD00038225
CAS 22398-14-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if swallowed.