Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl methoxymethylenemalonate, 98+%

$55.55 - $480.26

Chemical Identifiers

Chemical Name or Material Dimethyl methoxymethylenemalonate
CAS 22398-14-7
Molecular Formula C7H10O5
Formula Weight 174.16
MDL Number MFCD00038225
Synonym Methoxymethylenemalonic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2380330

 
alfa aesar™
B2380330
250g Each for $480.26

AAB2380309

 
alfa aesar™
B2380309
10g Each for $55.55
Description & Specifications

Specifications

Formula Weight 174.16
Chemical Name or Material Dimethyl methoxymethylenemalonate
Flash Point 158°C (316°F)
Health Hazard 2 GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Molecular Formula C7H10O5
Melting Point 41° to 45°C
Boiling Point 97° to 99°C (0.5mmHg)
MDL Number MFCD00038225
CAS 22398-14-7
Quantity 250g
Odor Ester-like
Synonym Methoxymethylenemalonic acid dimethyl ester
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.