missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl Methoxymalonate, TCI America™

$123.75 - $433.89

Chemical Identifiers

CAS 5018-30-4
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00009847
InChI Key ORXJMBXYSGGCHG-UHFFFAOYSA-N
Synonym Methoxymalonic Acid Dimethyl Ester
PubChem CID 78718
IUPAC Name dimethyl 2-methoxypropanedioate
SMILES COC(C(=O)OC)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M08365G View Documents TCI America
M08365G
5g Each for $123.75
 
M083625G View Documents TCI America
M083625G
25g Each for $433.89
 
Specifications

Chemical Identifiers

5018-30-4
162.141
ORXJMBXYSGGCHG-UHFFFAOYSA-N
78718
COC(C(=O)OC)C(=O)OC
C6H10O5
MFCD00009847
Methoxymalonic Acid Dimethyl Ester
dimethyl 2-methoxypropanedioate

Specifications

110°C
5g
C6H10O5
Methoxymalonic Acid Dimethyl Ester
COC(C(=O)OC)C(=O)OC
162.141
162.14
≥95.0% (GC)
Yellow
5018-30-4
MFCD00009847
ORXJMBXYSGGCHG-UHFFFAOYSA-N
dimethyl 2-methoxypropanedioate
78718
Clear Liquid at 20°C
Dimethyl Methoxymalonate
Safety and Handling

Safety and Handling

TSCA : No