Learn More
Dimethyl malonate, 98+%, Thermo Scientific Chemicals
$52.05 - $323.84
Chemical Identifiers
CAS | 108-59-8 |
---|---|
Molecular Formula | C5H8O4 |
Molecular Weight (g/mol) | 132.115 |
MDL Number | MFCD00008460 |
InChI Key | BEPAFCGSDWSTEL-UHFFFAOYSA-N |
Synonym | dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester |
PubChem CID | 7943 |
IUPAC Name | dimethyl propanedioate |
SMILES | COC(=O)CC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1100722
|
Thermo Scientific Chemicals
A1100722 |
100 g |
Each for $52.05
|
|
|||||
AAA1100736
|
Thermo Scientific Chemicals
A1100736 |
500 g |
Each for $90.75
|
|
|||||
AAA110070E
|
Thermo Scientific Chemicals
A110070E |
2500 g |
Each for $323.84
|
|
|||||
Description
Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.
Solubility
Miscible with alcohol, ether and oils. Slightly miscible with water.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Chemical Identifiers
108-59-8 | |
132.115 | |
BEPAFCGSDWSTEL-UHFFFAOYSA-N | |
7943 | |
COC(=O)CC(=O)OC |
C5H8O4 | |
MFCD00008460 | |
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester | |
dimethyl propanedioate |
Specifications
108-59-8 | |
1.156 | |
90°C (194°F) | |
1.413 | |
100 g | |
14,6096 | |
Miscible with alcohol,ether and oils. Slightly miscible with water. | |
COC(=O)CC(=O)OC | |
132.115 | |
132.12 | |
Dimethyl malonate |
-62°C | |
180°C to 181°C | |
C5H8O4 | |
MFCD00008460 | |
774261 | |
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester | |
BEPAFCGSDWSTEL-UHFFFAOYSA-N | |
dimethyl propanedioate | |
7943 | |
≥98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P233-P240-P241-P242-P243-P264b-P280-P305+P351+P338-P374-P501c
H227-H320
EINECSNumber : 203-597-8
RTECSNumber : OO0950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only