missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl Maleate, TCI America™

Manufacturer:  TCI America M001225ML

 View more versions of this product

Catalog No. M001225ML



Specifications

Specifications

Dimethyl Maleate
Yellow
25mL
C6H8O4
2810
LDCRTTXIJACKKU-ARJAWSKDSA-N
dimethyl (Z)-but-2-enedioate
5271565
144.13
≥97.0% (GC)
205°C
-19°C
624-48-6
MFCD00008459
dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z
COC(=O)C=CC(=O)OC
144.126
CHEBI:35460
Clear Liquid at 20°C

Chemical Identifiers

624-48-6
144.126
LDCRTTXIJACKKU-ARJAWSKDSA-N
5271565
dimethyl (Z)-but-2-enedioate
C6H8O4
MFCD00008459
dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z
CHEBI:35460
COC(=O)C=CC(=O)OC
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1107&(2)-1142

RTECSNumber : EM6300000

TSCA : Yes

Documents