Learn More
Dimethyl maleate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 218460010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethyl maleate | |
624-48-6 | |
1.1500g/mL | |
95°C | |
95% min. (GC) | |
C6H8O4 | |
CH3O2CCH=CHCO2CH3 | |
1 L | |
1.15 | |
Solubility in water: 80g/L (25°C) | |
COC(=O)C=CC(=O)OC | |
144.13 | |
4 mPa.s (20°C) | |
144.13 | |
Liquid |
96% | |
-17.5°C | |
204.0°C to 205.0°C | |
Authentic | |
Glass bottle | |
1.4410 to 1.4430 | |
MFCD00008459 | |
02,751 | |
dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z | |
LDCRTTXIJACKKU-ARJAWSKDSA-N | |
dimethyl (Z)-but-2-enedioate | |
5271565 | |
CHEBI:35460 | |
96% |
Chemical Identifiers
624-48-6 | |
144.13 | |
LDCRTTXIJACKKU-ARJAWSKDSA-N | |
5271565 | |
dimethyl (Z)-but-2-enedioate |
C6H8O4 | |
MFCD00008459 | |
dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z | |
CHEBI:35460 | |
COC(=O)C=CC(=O)OC |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Danger
EINECSNumber : 210-848-5
RTECSNumber : EM6300000
TSCA : TSCA
RUO – Research Use Only