missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl maleate, 96%

$32.47 - $153.78

Chemical Identifiers

CAS 624-48-6
Molecular Formula C6H8O4
Formula Weight 144.13
MDL Number MFCD00008459
UN Number UN2810
Beilstein 471705
Synonym Maleic acid dimethyl ester; cis-Propenoic acid dimethyl ester
Chemical Name or Material Dimethyl maleate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA189440B SDS Alfa Aesar™
A189440B
1000g Each for $153.78
AAA1894422 SDS Alfa Aesar™
A1894422
100g Each for $32.47
AAA1894430 SDS Alfa Aesar™
A1894430
250g Each for $49.57
AAA189440I SDS Alfa Aesar™
A189440I
5000g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.

Solubility
Miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Description & Specifications

Chemical Identifiers

624-48-6
144.13
UN2810
Maleic acid dimethyl ester; cis-Propenoic acid dimethyl ester
C6H8O4
MFCD00008459
471705
Dimethyl maleate

Specifications

91°C (195°F)
−19°C
624-48-6
144.13
UN2810
Maleic acid dimethyl ester; cis-Propenoic acid dimethyl ester
96%
204° to 205°C
1000g
C6H8O4
MFCD00008459
471705
Dimethyl maleate

Safety and Handling

GHS H Statement
H311-H318-H227-H302-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : EM6300000

TSCA : TSCA