missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl L-tartrate, 99%, Thermo Scientific Chemicals
$55.97 - $140.04
Chemical Identifiers
CAS | 608-68-4 |
---|---|
Molecular Formula | C6H10O6 |
Molecular Weight (g/mol) | 178.14 |
MDL Number | MFCD00064437 |
InChI Key | PVRATXCXJDHJJN-QWWZWVQMSA-N |
Synonym | +-dimethyl l-tartrate, dimethyl l-tartrate, 2r,3r-dimethyl 2,3-dihydroxysuccinate, dimethyl +-tartrate, l-+-tartaric acid dimethyl ester, dimethyl l-+-tartrate, 1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester, dimethyl 2r,3r-2,3-dihydroxybutanedioate, l-tartaric acid, dimethyl ester |
PubChem CID | 11851 |
IUPAC Name | dimethyl (2R,3R)-2,3-dihydroxybutanedioate |
SMILES | COC(=O)C(C(C(=O)OC)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0656114
|
Thermo Scientific Chemicals
L0656114 |
25 g |
Each for $55.97
|
|
|||||
AAL0656122
|
Thermo Scientific Chemicals
L0656122 |
100 g |
Each for $140.04
|
|
|||||
Description
Dimethyl L-tartrate is a chiral auxiliary used for the enantioselective epoxidation of allylic alcohols. It is used for asymmetric α-halogenation of acetals. It is a starting material for the synthesis of chiral building blocks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl L-tartrate is a chiral auxiliary used for the enantioselective epoxidation of allylic alcohols. It is used for asymmetric α-halogenation of acetals. It is a starting material for the synthesis of chiral building blocks.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
608-68-4 | |
178.14 | |
PVRATXCXJDHJJN-QWWZWVQMSA-N | |
11851 | |
COC(=O)C(C(C(=O)OC)O)O |
C6H10O6 | |
MFCD00064437 | |
+-dimethyl l-tartrate, dimethyl l-tartrate, 2r,3r-dimethyl 2,3-dihydroxysuccinate, dimethyl +-tartrate, l-+-tartaric acid dimethyl ester, dimethyl l-+-tartrate, 1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester, dimethyl 2r,3r-2,3-dihydroxybutanedioate, l-tartaric acid, dimethyl ester | |
dimethyl (2R,3R)-2,3-dihydroxybutanedioate |
Specifications
608-68-4 | |
1.3 | |
>170°C (338°F) | |
MFCD00064437 | |
1726256 | |
Soluble in water. | |
+21° (c=1 in Water) | |
dimethyl (2R,3R)-2,3-dihydroxybutanedioate | |
11851 | |
99% |
56°C to 61°C | |
158°C (11 mmHg) | |
C6H10O6 | |
25 g | |
+-dimethyl l-tartrate, dimethyl l-tartrate, 2r,3r-dimethyl 2,3-dihydroxysuccinate, dimethyl +-tartrate, l-+-tartaric acid dimethyl ester, dimethyl l-+-tartrate, 1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester, dimethyl 2r,3r-2,3-dihydroxybutanedioate, l-tartaric acid, dimethyl ester | |
PVRATXCXJDHJJN-QWWZWVQMSA-N | |
COC(=O)C(C(C(=O)OC)O)O | |
178.14 | |
178.14 | |
Dimethyl L-tartrate |
Safety and Handling
EINECSNumber : 210-166-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only