Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl isopropenyl phosphate, 95%

$40.03 - $317.09

Chemical Identifiers

Chemical Name or Material Dimethyl isopropenyl phosphate
CAS 4185-82-4
Molecular Formula C5H11O4P
Formula Weight 166.11
MDL Number MFCD09953485
Synonym Dimethyl 1-methylethenyl phosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2735306

 
alfa aesar™
H2735306
5g Each for $40.03

AAH2735314

 
alfa aesar™
H2735314
25g Each for $129.92

AAH2735322

 
alfa aesar™
H2735322
100g Each for $317.09
Description & Specifications

Specifications

Formula Weight 166.11
Chemical Name or Material Dimethyl isopropenyl phosphate
Assay 95%
Molecular Formula C5H11O4P
Boiling Point 120°C (7.5mmHg)
MDL Number MFCD09953485
CAS 4185-82-4
Quantity 5g
Synonym Dimethyl 1-methylethenyl phosphate