Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl ethylphosphonate, 97%

$310.39 - $1,226.50

Chemical Identifiers

Chemical Name or Material Dimethyl ethylphosphonate
CAS 6163-75-3
Molecular Formula C4H11O3P
Formula Weight 138.1
MDL Number MFCD00065120
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4464206

  SDS
alfa aesar™
4464206
5g Each for $310.39

AA4464214

  SDS
alfa aesar™
4464214
25g Each for $1,226.50
Description & Specifications

Specifications

Formula Weight 138.1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C4H11O3P
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 61° to 62°C (6mmHg)
CAS 6163-75-3
Quantity 5g
Color Colorless
MDL Number MFCD00065120
Physical Form Liquid
Assay 97%
Chemical Name or Material Dimethyl ethylphosphonate
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.