Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl(dimethylamino)vinylsilane, 97%

Chemical Identifiers

Chemical Name or Material Dimethyl(dimethylamino)vinylsilane
CAS 13391-72-5
Molecular Formula C6H15NSi
Formula Weight 129.28
MDL Number MFCD01318143
UN Number UN2924
Beilstein 2203481
Synonym (Dimethylamino)dimethylvinylsilane; (Dimethylamino)vinyldimethylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1728406

  SDS
alfa aesar™
L1728406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.28
Chemical Name or Material Dimethyl(dimethylamino)vinylsilane
CAS 13391-72-5
Health Hazard 1 Danger
MDL Number MFCD01318143
Assay 97%
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN2924
Synonym (Dimethylamino)dimethylvinylsilane; (Dimethylamino)vinyldimethylsilane
Molecular Formula C6H15NSi
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Beilstein 2203481
Recommended Storage 2° to 8°C
Boiling Point 109°C
Quantity 5g
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C