Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl cyclohexylphosphonate

$40.47 - $404.82

Chemical Identifiers

Chemical Name or Material Dimethyl cyclohexylphosphonate
CAS 1641-61-8
Molecular Formula C6H11P(O)(OCH3)2
Formula Weight 193.39
MDL Number MFCD00014300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4436409

  SDS
alfa aesar™
4436409
10g Each for $404.82

AA4436402

  SDS
alfa aesar™
4436402
0.5g Each for $40.47

AA4436404

  SDS
alfa aesar™
4436404
2g Each for $105.89
Description & Specifications

Specifications

Formula Weight 193.39
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1641-61-8
Health Hazard 1 Warning
Chemical Name or Material Dimethyl cyclohexylphosphonate
Physical Form Liquid
Molecular Formula C6H11P(O)(OCH3)2
Boiling Point 30° to 32°C (0.1mmHg)
MDL Number MFCD00014300
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.