missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl cyclohexylphosphonate

Manufacturer:  Alfa Aesar™ 4436404

 View more versions of this product

Catalog No. AA4436404



Specifications

Specifications

Dimethyl cyclohexylphosphonate
2g
C8H17O3P
acmc-1byde
COP(=O)(C1CCCCC1)OC
192.195
193.39
30°C to 32°C (0.1mmHg)
1641-61-8
MFCD00014300
OZOVYDFZXJIQPR-UHFFFAOYSA-N
dimethoxyphosphorylcyclohexane
12589506
Liquid

Chemical Identifiers

1641-61-8
192.195
OZOVYDFZXJIQPR-UHFFFAOYSA-N
12589506
COP(=O)(C1CCCCC1)OC
C8H17O3P
MFCD00014300
acmc-1byde
dimethoxyphosphorylcyclohexane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents