Please login to your online account to display your discounted pricing

Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, 96%, ACROS Organics™


Quantity 100mg

Manufacturer: thermo fisher scientific  235531000

 View more versions of this product

Catalog No. AC235531000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95% methylated beta-cyclodextrins (calculated), 40 to 60% isomer purity (heptakis(2,6-di-O-methyl)-beta-cyclodextrin), 0.5% max. unmodified beta-cyclodextrin
Physical Form Crystalline Powder
Formula Weight 1331.4
Quantity 100mg
Color White to faintly yellow
Water 5% Max. (K.F.)
Specific Rotation Condition +163° (20°C c=1,water)
Chemical Name or Material Dimethyl-β-cyclodextrin, methylated β-cyclodextrins
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C56H98O35
MDL Number MFCD00011616
CAS 51166-71-3
Synonym DIMEB
NMR 12.0 to 16.0 methyl groups per cyclodextrin ring (average degree of substitution)

Specifications

Chemical Name or Material Dimethyl-β-cyclodextrin, methylated β-cyclodextrins
CAS 51166-71-3
Assay ≥95% methylated beta-cyclodextrins (calculated), 40 to 60% isomer purity (heptakis(2,6-di-O-methyl)-beta-cyclodextrin), 0.5% max. unmodified beta-cyclodextrin
Assay Percent Range 96%
Molecular Formula C56H98O35
Formula Weight 1331.4
MDL Number MFCD00011616
Synonym DIMEB