Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl cis-4-cyclohexene-1,2-dicarboxylate, 99%

$44.59 - $192.86

Chemical Identifiers

Chemical Name or Material Dimethyl cis-4-cyclohexene-1,2-dicarboxylate
CAS 4841-84-3
Molecular Formula C10H14O4
Formula Weight 198.22
MDL Number MFCD00209581
Synonym cis-4-Cyclohexene-1,2-dicarboxylic acid dimethyl ester; Dimethyl cis-1,2,3,6-tetrahydrophthalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2595203

  SDS
alfa aesar™
H2595203
1g Each for $44.59

AAH2595209

  SDS
alfa aesar™
H2595209
10g Each for $192.86
Description & Specifications

Specifications

Formula Weight 198.22
Synonym cis-4-Cyclohexene-1,2-dicarboxylic acid dimethyl ester; Dimethyl cis-1,2,3,6-tetrahydrophthalate
Assay 99%
Quantity 1g
MDL Number MFCD00209581
Flash Point >110°C (230°F)
Boiling Point 141° to 142°C (20mmHg)
Molecular Formula C10H14O4
Chemical Name or Material Dimethyl cis-4-cyclohexene-1,2-dicarboxylate
CAS 4841-84-3