Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl biphenyl-4,4'-dicarboxylate, 98%

$76.65 - $1,146.77

Chemical Identifiers

Chemical Name or Material Dimethyl biphenyl-4,4'-dicarboxylate
CAS 792-74-5
Molecular Formula C16H14O4
Formula Weight 270.29
MDL Number MFCD00017201
Beilstein 2055852
Synonym Biphenyl-4'4-dicarboxylic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1255603

 
alfa aesar™
A1255603
1g Each for $76.65

AAA1255606

 
alfa aesar™
A1255606
5g Each for $290.92

AAA1255614

 
alfa aesar™
A1255614
25g Each for $1,146.77
Description & Specifications

Specifications

Formula Weight 270.29
Chemical Name or Material Dimethyl biphenyl-4,4'-dicarboxylate
TSCA TSCA
Assay 98%
Molecular Formula C16H14O4
Melting Point 212° to 216°C
MDL Number MFCD00017201
CAS 792-74-5
Beilstein 2055852
Quantity 1g
Synonym Biphenyl-4'4-dicarboxylic acid dimethyl ester

Applications
Dimethyl biphenyl-4,4′-dicarboxylate is used as a hepatoprotectant in the treatment of chronic hepatitis. At 250 µg/ml, it has been shown to stimulate JAK/STAT signaling and to induce the expression of interferon-α stimulated genes in a HepG2 cell line. It has been used to study hepatic fibrosis and chronic hepatocellular damage. It appears to normalize alanine aminotransferase (ALT) in cases where levels of ALT are elevated. It may be employed as starting reagent for the synthesis of dimethyl 2-fluoro-and 2,2′-difluorobiphenyl-4,4′-dicarboxylates.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.