Please login to your online account to display your discounted pricing

Dimethyl aminoterephthalate, 99%, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  194780500

 View more versions of this product

Catalog No. AC194780500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 209.2
Color Beige or yellow to light green to brownish
Physical Form Powder or Crystals
Quantity 50g
TSCA TSCA
EINECS Number 226-364-2
Melting Point 131° to 134°C
Beilstein 14, 559
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Plastic bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Dimethyl aminoterephthalate
MDL Number MFCD00008427
Name Note 99%
Linear Formula H2NC6H3(CO2CH3)2
Molecular Formula C10H11NO4
Assay Percent Range 99%
CAS 5372-81-6

Specifications

Chemical Name or Material Dimethyl aminoterephthalate
Name Note 99%
CAS 5372-81-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H11NO4
Linear Formula H2NC6H3(CO2CH3)2
Formula Weight 209.2
MDL Number MFCD00008427
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents