missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1056730

 View more versions of this product

Catalog No. AAA1056730



Description

Description

Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Dimethyl aminoterephthalate
130°C to 134°C
MFCD00008427
2808505
Insoluble in water.
COC(=O)C1=CC(=C(C=C1)C(=O)OC)N
209.201
209.2
5372-81-6
C10H11NO4
250 g
dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester
DSSKDXUDARIMTR-UHFFFAOYSA-N
dimethyl 2-aminobenzene-1,4-dicarboxylate
79336
99%

Chemical Identifiers

5372-81-6
209.201
DSSKDXUDARIMTR-UHFFFAOYSA-N
79336
COC(=O)C1=CC(=C(C=C1)C(=O)OC)N
C10H11NO4
MFCD00008427
dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester
dimethyl 2-aminobenzene-1,4-dicarboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-364-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only