missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl aminomalonate hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 374970250

 View more versions of this product

Catalog No. AC374970250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Dimethyl aminomalonate hydrochloride
Authentic
Glass bottle
H2NCH(CO2CH3)2·HCl
25g
QWNDKNJSEWOEDM-UHFFFAOYSA-N
dimethyl 2-aminopropanedioate;hydrochloride
2734846
97%
16115-80-3
96% min. (Argentometry)
C5H10ClNO4
MFCD00038880
dimethyl 2-aminomalonate hydrochloride, dimethyl aminomalonate hydrochloride, dimethyl aminomalonate hcl, aminomalonic acid dimethyl ester hydrochloride, 1,3-dimethyl 2-aminopropanedioate hydrochloride, dimethyl 2-aminopropanedioate hydrochloride, acmc-1bp9z, 2-amino-malonic acid dimethyl ester hydrochloride, 2-aminomalonic acid dimethyl ester hydrochloride
[H+].[Cl-].COC(=O)C(N)C(=O)OC
183.59
183.59

Chemical Identifiers

16115-80-3
183.59
QWNDKNJSEWOEDM-UHFFFAOYSA-N
2734846
[H+].[Cl-].COC(=O)C(N)C(=O)OC
C5H10ClNO4
MFCD00038880
dimethyl 2-aminomalonate hydrochloride, dimethyl aminomalonate hydrochloride, dimethyl aminomalonate hcl, aminomalonic acid dimethyl ester hydrochloride, 1,3-dimethyl 2-aminopropanedioate hydrochloride, dimethyl 2-aminopropanedioate hydrochloride, acmc-1bp9z, 2-amino-malonic acid dimethyl ester hydrochloride, 2-aminomalonic acid dimethyl ester hydrochloride
dimethyl 2-aminopropanedioate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 240-284-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only