missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl allylphosphonate, tech. 85%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1056706

 View more versions of this product

Catalog No. AAL1056706



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethyl allylphosphonate
41°C to 42°C (0.5 mmHg)
1.437
5 g
dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester
COP(=O)(CC=C)OC
150.114
150.12
Technical
757-54-0
C5H11O3P
MFCD00014951
1756526
ZOSQAGGCVFVCNO-UHFFFAOYSA-N
3-dimethoxyphosphorylprop-1-ene
136570
85%

Chemical Identifiers

757-54-0
150.114
ZOSQAGGCVFVCNO-UHFFFAOYSA-N
136570
COP(=O)(CC=C)OC
C5H11O3P
MFCD00014951
dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester
3-dimethoxyphosphorylprop-1-ene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only