Please login to your online account to display your discounted pricing

Dimethyl allylphosphonate, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  330680010

 View more versions of this product

Catalog No. AC330680010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 150.11
Quantity 1g
Color Clear colorless
Refractive Index 1.4360 to 1.4400 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00014951
Assay Percent Range 97%
Molecular Formula C5H11O3P
Chemical Name or Material Dimethyl allylphosphonate
Name Note 97%
CAS 757-54-0

Specifications

Chemical Name or Material Dimethyl allylphosphonate
Name Note 97%
CAS 757-54-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H11O3P
Formula Weight 150.11
MDL Number MFCD00014951