Please login to your online account to display your discounted pricing

Dimethyl allylphosphonate, 97%, ACROS Organics™

$39.71 - $39.71

Chemical Identifiers

Chemical Name or Material Dimethyl allylphosphonate
Name Note 97%
CAS 757-54-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H11O3P
Formula Weight 150.11
MDL Number MFCD00014951
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC330680010

  SDS
thermo fisher scientific
330680010
Glass bottle 1g Each for $39.71
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 150.11
Quantity 1g
Color Clear colorless
Refractive Index 1.4360 to 1.4400 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00014951
Assay Percent Range 97%
Molecular Formula C5H11O3P
Chemical Name or Material Dimethyl allylphosphonate
Name Note 97%
CAS 757-54-0

for C-C Bond Formation (Olefination)