Learn More
Dimethyl adipimidate dihydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1051509
Description
Dimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane
Solubility
Hydrolyses in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, water.
Specifications
Dimethyl adipimidate dihydrochloride | |
∽215°C (decomposition) | |
MFCD00012573 | |
Moisture sensitive | |
Hydrolyses in water. | |
COC(=N)CCCCC(=N)OC.Cl.Cl | |
245.144 | |
245.15 |
14620-72-5 | |
C8H18Cl2N2O2 | |
10 g | |
dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride | |
IEUUDEWWMRQUDS-UHFFFAOYSA-N | |
dimethyl hexanediimidate;dihydrochloride | |
12876176 | |
97% |
Chemical Identifiers
14620-72-5 | |
245.144 | |
IEUUDEWWMRQUDS-UHFFFAOYSA-N | |
12876176 | |
COC(=N)CCCCC(=N)OC.Cl.Cl |
C8H18Cl2N2O2 | |
MFCD00012573 | |
dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride | |
dimethyl hexanediimidate;dihydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-660-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only