missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl adipate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 181315000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethyl adipate | |
627-93-0 | |
8.0°C | |
109.0°C to 110.0°C (14.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4270 to 1.4290 | |
MFCD00008469 | |
02, 652 | |
10, 151 | |
Solubility in water: insoluble. Other solubilities: soluble in ether,alcohols,carbon tetrachloride,,acetic acid | |
COC(=O)CCCCC(=O)OC | |
174.2 | |
5 mPa.s (20°C) | |
174.2 | |
Liquid |
99+% | |
0.01% max. (as adipic acid) | |
1.0600g/mL | |
107°C | |
99% min. (GC) | |
C8H14O4 | |
CH3O2C(CH2)4CO2CH3 | |
500 g | |
1.06 | |
dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62 | |
UDSFAEKRVUSQDD-UHFFFAOYSA-N | |
dimethyl hexanedioate | |
12329 | |
CHEBI:34715 | |
99+% |
Chemical Identifiers
627-93-0 | |
174.2 | |
UDSFAEKRVUSQDD-UHFFFAOYSA-N | |
12329 | |
dimethyl hexanedioate |
C8H14O4 | |
MFCD00008469 | |
dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62 | |
CHEBI:34715 | |
COC(=O)CCCCC(=O)OC |
Safety and Handling
EINECSNumber : 211-020-6
RTECSNumber : AV1645000
TSCA : TSCA
RUO – Research Use Only