missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0053714
Description
Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.
Solubility
Difficult to mix.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Dimethyl acetylmethylphosphonate | |
1.202 | |
93°C (199°F) | |
1.439 | |
25 g | |
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone | |
UOWIYNWMROWVDG-UHFFFAOYSA-N | |
1-dimethoxyphosphorylpropan-2-one | |
77872 | |
97% |
4202-14-6 | |
121°C to 123°C (11 mmHg) | |
C5H11O4P | |
MFCD00008769 | |
1766218 | |
Difficult to mix. | |
CC(=O)CP(=O)(OC)OC | |
166.113 | |
166.12 |
Chemical Identifiers
4202-14-6 | |
166.113 | |
UOWIYNWMROWVDG-UHFFFAOYSA-N | |
77872 | |
CC(=O)CP(=O)(OC)OC |
C5H11O4P | |
MFCD00008769 | |
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone | |
1-dimethoxyphosphorylpropan-2-one |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 224-110-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only