Learn More
Dimethyl acetylenedicarboxylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1143736
Description
It acts as an inhibitor of oxidative phosphorylation. It is also used in Pharmaceutical and electronic industry. It is used in organic synthesis. It also acts as an intermediate in chemical research. It is also used in Diels-Alder Reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt acts as an inhibitor of oxidative phosphorylation. It is also used in Pharmaceutical and electronic industry. It is used in organic synthesis. It also acts as an intermediate in chemical research. It is also used in Diels-Alder Reaction.
Solubility
It is insoluble in water but soluble in ethanol, ethyl ether, CCl4, miscible with organic solvents.
Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents, strong acids, strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Dimethyl acetylenedicarboxylate | |
-18°C | |
95°C to 98°C (19 mmHg) | |
C6H6O4 | |
CH3O2CC≡CCO2CH3 | |
500 g | |
607063 | |
dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene | |
VHILMKFSCRWWIJ-UHFFFAOYSA-N | |
dimethyl but-2-ynedioate | |
12980 | |
98% |
762-42-5 | |
1.156 | |
86°C (186°F) | |
1.448 | |
MFCD00008456 | |
UN1760 | |
Light sensitive | |
It is insoluble in water but soluble in ethanol,ethyl ether,CCl4,miscible with organic solvents. | |
COC(=O)C#CC(=O)OC | |
142.11 | |
142.11 |
Chemical Identifiers
762-42-5 | |
142.11 | |
VHILMKFSCRWWIJ-UHFFFAOYSA-N | |
12980 | |
COC(=O)C#CC(=O)OC |
C6H6O4 | |
MFCD00008456 | |
dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene | |
dimethyl but-2-ynedioate |
Safety and Handling
GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber : 212-098-4
RTECSNumber : ES0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only