missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl acetone-1,3-dicarboxylate, 97%

Manufacturer:  Alfa Aesar™ A1496936

 View more versions of this product

Catalog No. AAA1496936



Specifications

Specifications

Dimethyl acetone-1,3-dicarboxylate
Ester-like
16°C to 17°C
1830-54-2
MFCD00008462
dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
COC(=O)CC(=O)CC(=O)OC
174.152
174.16
>110°C (230°F)
237°C to 241°C
500g
C7H10O5
1366042
RNJOKCPFLQMDEC-UHFFFAOYSA-N
dimethyl 3-oxopentanedioate
74591
97%

Chemical Identifiers

1830-54-2
174.152
RNJOKCPFLQMDEC-UHFFFAOYSA-N
74591
COC(=O)CC(=O)CC(=O)OC
C7H10O5
MFCD00008462
dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
dimethyl 3-oxopentanedioate
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P280g-P262
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents