Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl 5-sulfoisophthalate sodium salt, 98%

$34.01 - $306.31

Chemical Identifiers

Chemical Name or Material Dimethyl 5-sulfoisophthalate sodium salt
CAS 3965-55-7
Molecular Formula C10H9NaO7S
Formula Weight 296.23
MDL Number MFCD00007493
Synonym 5-Sulfoisophthalic acid dimethyl ester sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2464614

  SDS
alfa aesar™
B2464614
25g Each for $34.01

AAB2464622

  SDS
alfa aesar™
B2464622
100g Each for $91.73

AAB2464636

  SDS
alfa aesar™
B2464636
500g Each for $306.31
Description & Specifications

Specifications

Formula Weight 296.23
Chemical Name or Material Dimethyl 5-sulfoisophthalate sodium salt
TSCA TSCA
Assay 98%
Molecular Formula C10H9NaO7S
Melting Point >300°C
MDL Number MFCD00007493
CAS 3965-55-7
Quantity 25g
Synonym 5-Sulfoisophthalic acid dimethyl ester sodium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.