missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl 5-bromoisophthalate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5062903

 View more versions of this product

Catalog No. AAH5062903


Description

Description

Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Dimethyl 5-bromoisophthalate
88°C to 90°C
MFCD00078709
dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
QUJINGKSNJNXEB-UHFFFAOYSA-N
dimethyl 5-bromobenzene-1,3-dicarboxylate
103954
98%
51760-21-5
C10H9BrO4
1 g
Insoluble in water.
COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC
273.082
273.08

Chemical Identifiers

51760-21-5
273.082
QUJINGKSNJNXEB-UHFFFAOYSA-N
103954
COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC
C10H9BrO4
MFCD00078709
dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
dimethyl 5-bromobenzene-1,3-dicarboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 257-386-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only