Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl 4-nitrophthalate, 98%

$28.93 - $28.93

Chemical Identifiers

Chemical Name or Material Dimethyl 4-nitrophthalate
CAS 610-22-0
Molecular Formula C10H9NO6
Formula Weight 239.19
MDL Number MFCD00017191
Beilstein 1886234
Synonym Dimethyl 4-nitrobenzene-1,2-dicarboxylate; 4-Nitrophthalic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1788503

  SDS
alfa aesar™
A1788503
1g Each for $28.93
Description & Specifications

Specifications

Formula Weight 239.19
Chemical Name or Material Dimethyl 4-nitrophthalate
TSCA TSCA
Molecular Formula C10H9NO6
Boiling Point 192° to 195°C (25mmHg)
MDL Number MFCD00017191
Synonym Dimethyl 4-nitrobenzene-1,2-dicarboxylate; 4-Nitrophthalic acid dimethyl ester
Assay 98%
Melting Point 65° to 68°C
CAS 610-22-0
Beilstein 1886234
Quantity 1g