Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl (4-methoxybenzyl)malonate, 95%

$100.36 - $326.32

Chemical Identifiers

Chemical Name or Material Dimethyl (4-methoxybenzyl)malonate
CAS 15378-09-3
Molecular Formula C13H16O5
Formula Weight 252.27
MDL Number MFCD08436113
Synonym (4-Methoxybenzyl)malonic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2633103

 
alfa aesar™
H2633103
1g Each for $100.36

AAH2633106

 
alfa aesar™
H2633106
5g Each for $326.32
Description & Specifications

Specifications

Formula Weight 252.27
Chemical Name or Material Dimethyl (4-methoxybenzyl)malonate
Assay 95%
Molecular Formula C13H16O5
MDL Number MFCD08436113
Quantity 1g
Synonym (4-Methoxybenzyl)malonic acid dimethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
CAS 15378-09-3
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.