Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl (2-thienylmethyl)malonate, 95%

$156.70 - $391.76

Chemical Identifiers

Chemical Name or Material Dimethyl (2-thienylmethyl)malonate
CAS 122308-25-2
Molecular Formula C10H12O4S
Formula Weight 228.27
MDL Number MFCD08436111
Synonym (2-Thienylmethyl)malonic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2630103

  SDS
alfa aesar™
H2630103
1g Each for $391.76

AAH26301MD

  SDS
alfa aesar™
H26301MD
250mg Each for $156.70
Description & Specifications

Specifications

Formula Weight 228.27
Chemical Name or Material Dimethyl (2-thienylmethyl)malonate
Assay 95%
Molecular Formula C10H12O4S
MDL Number MFCD08436111
CAS 122308-25-2
Quantity 1g
Synonym (2-Thienylmethyl)malonic acid dimethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.