Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl 2-nitroterephthalate, 98+%

$39.88 - $494.11

Chemical Identifiers

Chemical Name or Material Dimethyl 2-nitroterephthalate
CAS 5292-45-5
Molecular Formula C10H9NO6
Formula Weight 239.19
MDL Number MFCD00008422
Beilstein 1993064
Synonym 2-Nitrobenzene-1,4-dicarboxylic acid dimethyl ester; 2-Nitroterephthalic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA129610B

 
alfa aesar™
A129610B
1000g Each for $494.11

AAA1296118

 
alfa aesar™
A1296118
50g Each for $39.88

AAA1296130

 
alfa aesar™
A1296130
250g Each for $147.31
Description & Specifications

Specifications

Formula Weight 239.19
Chemical Name or Material Dimethyl 2-nitroterephthalate
TSCA TSCA
Assay ≥98%
Molecular Formula C10H9NO6
Melting Point 72° to 75°C
MDL Number MFCD00008422
CAS 5292-45-5
Beilstein 1993064
Quantity 1000g
Synonym 2-Nitrobenzene-1,4-dicarboxylic acid dimethyl ester; 2-Nitroterephthalic acid dimethyl ester