Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl (2-methylbutyl)malonate, 97%

$89.80 - $224.51

Chemical Identifiers

Chemical Name or Material Dimethyl (2-methylbutyl)malonate
CAS 59107-99-2
Molecular Formula C10H18O4
Formula Weight 202.25
MDL Number MFCD08436117
Synonym (2-Methylbutyl)malonic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2658803

  SDS
alfa aesar™
H2658803
1g Each for $224.51

AAH26588MD

  SDS
alfa aesar™
H26588MD
250mg Each for $89.80
Description & Specifications

Specifications

Formula Weight 202.25
Chemical Name or Material Dimethyl (2-methylbutyl)malonate
Assay 97%
Synonym (2-Methylbutyl)malonic acid dimethyl ester
Molecular Formula C10H18O4
MDL Number MFCD08436117
CAS 59107-99-2
Quantity 1g