Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl 2-fluoroterephthalate, 98%

$95.54 - $95.54

Chemical Identifiers

Chemical Name or Material Dimethyl 2-fluoroterephthalate
CAS 5292-47-7
Molecular Formula C10H9FO4
Formula Weight 212.17
MDL Number MFCD00042266
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4384903

  SDS
alfa aesar™
4384903
1g Each for $95.54
Description & Specifications

Specifications

Formula Weight 212.17
Molecular Formula C10H9FO4
Physical Form Powder
Melting Point 81° to 83°C
Quantity 1g
CAS 5292-47-7
Assay 98%
Chemical Name or Material Dimethyl 2-fluoroterephthalate
MDL Number MFCD00042266