missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC258440500
Specifications
(+)-Dimethyl 2, 3-O-isopropylidene-D-tartrate | |
135°C | |
Glass bottle | |
1.15 | |
+ 54 | |
Colorless to Yellow | |
37031-30-4 | |
100.0 | |
C9H14O6 | |
+-dimethyl 2,3-o-isopropylidene-d-tartrate, +-dimethyl-2,3-o-isopropylidine-d-tartrate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, dimethyl +-2,3-o-isopropylidene-d-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, dimethyl--2,3-o-isopropylidene-l-tartrate, pubchem6765 | |
CC1(OC(C(O1)C(=O)OC)C(=O)OC)C | |
218.21 | |
218.21 | |
97% |
1.1500g/mL | |
Authentic | |
1.4386 to 1.4406 | |
+ 54.00 (20.00°C c=neat) | |
139°C (21.0 hPa) | |
50mL | |
96.0 | |
96% min. (GC) | |
MFCD00040504 | |
ROZOUYVVWUTPNG-WDSKDSINSA-N | |
dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate | |
688156 | |
Liquid |
Chemical Identifiers
37031-30-4 | |
218.21 | |
ROZOUYVVWUTPNG-WDSKDSINSA-N | |
688156 | |
CC1(OC(C(O1)C(=O)OC)C(=O)OC)C |
C9H14O6 | |
MFCD00040504 | |
+-dimethyl 2,3-o-isopropylidene-d-tartrate, +-dimethyl-2,3-o-isopropylidine-d-tartrate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, dimethyl +-2,3-o-isopropylidene-d-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, dimethyl--2,3-o-isopropylidene-l-tartrate, pubchem6765 | |
dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate |