missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl 1H-Imidazole-4,5-dicarboxylate 98.0+%, TCI America™

$189.28 - $367.03

Chemical Identifiers

CAS 3304-70-9
Molecular Formula C7H8N2O4
Molecular Weight (g/mol) 184.151
MDL Number MFCD00051338
InChI Key CUIWFAXEALIQJS-UHFFFAOYSA-N
Synonym dimethyl 4,5-imidazoledicarboxylate, 1h-imidazole-4,5-dicarboxylic acid dimethyl ester, dimethyl imidazole-4,5-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, dimethyl ester, 4,5-dimethyl 1h-imidazole-4,5-dicarboxylate, imidazole-4,5-dicarboxylic acid, dimethyl ester, dimethyl 4,5-imidazole-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, 4,5-dimethyl ester, imidazole-4, dimethyl ester, 1h-imidazole-4, dimethyl ester
PubChem CID 76810
IUPAC Name dimethyl 1H-imidazole-4,5-dicarboxylate
SMILES COC(=O)C1=C(N=CN1)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I045610G View Documents Tci America
I045610G
10g Each for $189.28
 
I045625G View Documents Tci America
I045625G
25g Each for $367.03
 
Specifications

Chemical Identifiers

3304-70-9
184.151
CUIWFAXEALIQJS-UHFFFAOYSA-N
76810
COC(=O)C1=C(N=CN1)C(=O)OC
C7H8N2O4
MFCD00051338
dimethyl 4,5-imidazoledicarboxylate, 1h-imidazole-4,5-dicarboxylic acid dimethyl ester, dimethyl imidazole-4,5-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, dimethyl ester, 4,5-dimethyl 1h-imidazole-4,5-dicarboxylate, imidazole-4,5-dicarboxylic acid, dimethyl ester, dimethyl 4,5-imidazole-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, 4,5-dimethyl ester, imidazole-4, dimethyl ester, 1h-imidazole-4, dimethyl ester
dimethyl 1H-imidazole-4,5-dicarboxylate

Specifications

White-Yellow
3304-70-9
MFCD00051338
CUIWFAXEALIQJS-UHFFFAOYSA-N
dimethyl 1H-imidazole-4,5-dicarboxylate
76810
Crystal-Powder at 20°C
Dimethyl 1H-Imidazole-4,5-dicarboxylate
10g
C7H8N2O4
dimethyl 4,5-imidazoledicarboxylate, 1h-imidazole-4,5-dicarboxylic acid dimethyl ester, dimethyl imidazole-4,5-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, dimethyl ester, 4,5-dimethyl 1h-imidazole-4,5-dicarboxylate, imidazole-4,5-dicarboxylic acid, dimethyl ester, dimethyl 4,5-imidazole-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, 4,5-dimethyl ester, imidazole-4, dimethyl ester, 1h-imidazole-4, dimethyl ester
COC(=O)C1=C(N=CN1)C(=O)OC
184.151
184.15
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6924

TSCA : Yes