Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl (1-ethylpropyl)malonate, 97%

$240.12 - $240.12

Chemical Identifiers

Chemical Name or Material Dimethyl (1-ethylpropyl)malonate
CAS 39520-19-9
Molecular Formula C10H18O4
Formula Weight 202.25
MDL Number MFCD08436116
Synonym Dimethyl (diethylmethyl)malonate; (1-Ethylpropyl)malonic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2670127

  SDS
alfa aesar™
H2670127
2.5g Each for $240.12
Description & Specifications

Specifications

Formula Weight 202.25
Chemical Name or Material Dimethyl (1-ethylpropyl)malonate
Assay 97%
Molecular Formula C10H18O4
MDL Number MFCD08436116
CAS 39520-19-9
Quantity 2.5g
Synonym Dimethyl (diethylmethyl)malonate; (1-Ethylpropyl)malonic acid dimethyl ester