Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl 1,4-cyclohexanedicarboxylate, cis + trans, 90+%

$45.86 - $343.26

Chemical Identifiers

Chemical Name or Material Dimethyl 1,4-cyclohexanedicarboxylate
CAS 94-60-0
Name Note cis + trans
Molecular Formula C10H16O4
Formula Weight 200.23
MDL Number MFCD00001460
Beilstein 1876703
Synonym Cyclohexane-1,4-dicarboxylic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1417106

 
alfa aesar™
A1417106
5g Each for $45.86

AAA1417114

 
alfa aesar™
A1417114
25g Each for $121.74

AAA1417122

 
alfa aesar™
A1417122
100g Each for $343.26
Description & Specifications

Specifications

Formula Weight 200.23
Chemical Name or Material Dimethyl 1,4-cyclohexanedicarboxylate
TSCA TSCA
Name Note cis + trans
Assay ≥90%
Molecular Formula C10H16O4
Boiling Point 129° to 131°C (9mmHg)
MDL Number MFCD00001460
CAS 94-60-0
Beilstein 1876703
Quantity 5g
Flash Point >110°C (230°F)
Synonym Cyclohexane-1,4-dicarboxylic acid dimethyl ester

Applications
Dimethyl 1,4-cyclohexanedicarboxylate, mixture of cis and trans may be used in chemical synthesis studies. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.