Please login to your online account to display your discounted pricing

Dimethyl 1,4-cyclohexanedicarboxylate, 90-95%, mixture of cis and trans, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dimethyl 1, 4-cyclohexanedicarboxylate
Name Note 90-95%
CAS 94-60-0
Assay ≥90%
Assay Percent Range 90 to 95%
Molecular Formula C10H16O4
Linear Formula C6H10(CO2CH3)2
Formula Weight 200.23
MDL Number MFCD00001460
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC297170050

  SDS
thermo fisher scientific
AC297170050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid After Melting
Assay ≥90%
Formula Weight 200.23
Quantity 5g
Color Clear colorless
EINECS Number 202-347-5
TSCA TSCA
Density 1.1000g/mL
Refractive Index 1.4560 to 1.4600 (20°C, 589nm)
Infrared Spectrum Authentic
Molecular Formula C10H16O4
Linear Formula C6H10(CO2CH3)2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Dimethyl 1, 4-cyclohexanedicarboxylate
Name Note 90-95%
Assay Percent Range 90 to 95%
MDL Number MFCD00001460
CAS 94-60-0
Safety and Handling

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.