missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl 1,4-cyclohexanedicarboxylate, 90-95%, mixture of cis and trans, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC297170050

 View more versions of this product

Catalog No. AC297170050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Dimethyl 1, 4-cyclohexanedicarboxylate
Authentic
5g
94-60-0
Liquid After Melting
90 to 95%
C6H10(CO2CH3)2
MFCD00001460
1.1000g/mL
Clear colorless
90-95%
1.4560 to 1.4600 (20°C, 589nm)
≥90%
C10H16O4
200.23

Chemical Identifiers

94-60-0
C10H16O4
200.23
Dimethyl 1, 4-cyclohexanedicarboxylate, 90-95%
90 to 95%
C6H10(CO2CH3)2
MFCD00001460
≥90%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-347-5

TSCA : TSCA